Nalidixic acid 50 mg/mL in water, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid

J67194-8EQ J67194-XF J67194-8EQ J67194-XF
AAJ67194-XFEA 76.48 USD
AAJ67194-XF AAJ67194-8EQ CAAAJ67194-XF CAAAJ67194-8EQ
Nalidixic acid 50 mg/mL in water, sterile filtered
Nalidixic acid

Nalidixic acid, 50 mg/ml in water, sterile-filtered

Formula: C₁₂H₁₂N₂O₃
MW: 232.24
Storage Temperature: Refrigerator
MDL Number: MFCD00006884
CAS Number: 389-08-2
EINECS: 206-864-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR