Comply™ Lead Free Steam Indicator Tape, 3M™
Supplier: Healthcare Products
MMM1322-12MM MMM 1322-24MM MMM 1322-18MM MMM 1322-12MM
56222-073CS 274.19 USD
56222-073 56614-625 89368-632 89368-636
Comply™ Lead Free Steam Indicator Tape, 3M™
Sterilization Indicators
This lead free indicator tape for steam sterilization consists of an adhesive, backing and chemical indicator stripes.
  • Lead free ink does not require hazardous waste disposal
  • Can be written on or labeled with pre-printed labels
  • Designed for use on all wraps
The adhesive is an aggressive, pressure sensitive designed to adhere to a variety of wraps, including untreated woven and disposable non‑woven, paper and paper/plastic wraps in order to secure the pack during steam sterilization. After sterilization, the tape is designed to remove easily and cleanly from untreated woven wraps. The backing is a beige colored crepe paper and provides the stretch needed for pack expansion during sterilization. The chemical indicator lines will show a visual color change from off‑white/tan to dark brown/black when exposed to a steam sterilization process.
Certifications: Conforms to ANSI/AAMII/ISO 11140-1:2005