Comply™ Bowie-Dick Plus Test Packs, 3M™
Supplier: Healthcare Products
MMM00135LF MMM1233LF
34010-105BX 188.91 USD
34010-105 75787-618 75787-620 75787-622
Comply™ Bowie-Dick Plus Test Packs, 3M™
Sterilization Indicators
Comply™ Bowie-Dick test packs are used to monitor the performance of vacuum-assisted 132 to 134 °C (270 to 273 °F) steam sterilizers.
  • Lead-free ink chemistry
  • Early warning feature helps to prevent unexpected failures
  • Easy-to-read test sheet clearly identifies failures and can be filed for record keeping
Test packs will reproducibly detect residual air in the sterilizer chamber due to air leaks, air removal failures, and inadequate vacuum.
Certifications: Complies to ANSI/AAMI/ISO 11140-5:2007 Class 2.