Anti-SUC1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-103-189S 200-103-189S
RL200-103-189SEA 148 USD
RL200-103-189S CA10036-766
Anti-SUC1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Invertase has been assayed against 1.0 µg of Invertase (Candida) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2500;
Do not add Sodium Azide!

Type: Primary
Antigen: SUC1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: C, Ida Species, C, Ida
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR