Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-E81 100-401-E81
RL100-401-E81EA 518 USD
CA100-401-E81 RL100-401-E81
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Histone H3 antibody has been tested for use in ELISA and western blot. For western blots expect a band of approximately 15.4 kDa in size corresponding tothe Histone 3 Protein. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:2000;

Type: Primary
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, chicken, dog, monkey, Xenopus laevis, Arabidopsis thaliana, Caenorhabditis elegans, Fruit fly
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR