Shipper, Combo Ambient/Frozen, Therapak®
Supplier: Therapak
37811
10029-278CS 88.15 USD
10029-278
Shipper, Combo Ambient/Frozen, Therapak®
Mailers and Shippers
System includes Aqui-Pak, 6 bay absorbent pouch, 95 kPa Biohazard Specimen Bag, 6x6" zip style bag, Medium 95kPa Canister, Vertical Combo Shipper Box, 6-Vial Prostate Biopsy Kit Box, Dry Ice Label, Biohazard Label, Double Up Arrow Label, UN3373 Biological Substance, Cat B Label, List of Contents Card, Small Foam Cooler.