Wood Gnawing Blocks, Certified, Bio-Serv
Supplier: Bio-Serv
K3511-300 K3512-175 K3513-100
89412-118EA 77.04 USD
89412-118 89412-120 89412-122 89412-119 89412-121 89412-123
Wood Gnawing Blocks, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of high quality, kiln dried, 100% natural hardwood
Provides the most natural form of gnawing enrichment. Cube shape reduces loss through cage floor. Autoclavable.
Certifications: Certified (Contaminant Screened).
Ordering information: Hardwood blocks are 100% natural wood. Therefore, characteristic variations in wood grain patterns and/or color will be observed in the box.