ActivArmr® 80-600 Cut resistant gloves, Ansell
Supplier: Ansell Healthcare
206411 206409 206410
47734-558CS 1970.01 USD
47734-558 47734-562 47734-564 47734-560
ActivArmr® 80-600 Cut resistant gloves, Ansell
Gloves
Boasting excellent cut resistance with comfort and a sure grip, the PowerFlex® 80-600 glove incorporates a 100% DuPont™ Kevlar® liner with a thick natural rubber latex coating.
  • Silicone-free
  • Superior dry grip
  • Exceptional resistance to cuts, abrasions, and punctures
These gloves feature a crinkle finish latex coating for superior dry grip for bottling and glass handling, refuse collection and sheet metal handling.
The ergonomically designed gloves provide unmatched fit and comfort, while reducing hand fatigue. An uncoated back allows air to circulate to keep hands cooler and drier, and the fully coated thumb provides added protection in this high-risk area. Wearing the right size glove is the best way to ensure that hand protection products are comfortable. PowerFlex gloves are available in several sizes for a comfortable and adjusted fit.
Use in the appliance manufacturing, automotive, glass handling, masonry, tile, and plumbing industries. Uses include sheet metal, glass handling, recycling, canning, and bottling.
The 80-600 allows user to focus on the job without worrying about bulky, uncomfortable cut-protection gloves. Thumb is fully coated for additional protection in this high-wear area. Ergonomically dipped for reduced hand fatigue with no uncomfortable seams.
Certifications: ANSI level 2 cut protection.
Packaging: Gloves are packaged 12 pairs in a bag; 72 pairs in a carton.