Rodent Huts, Certified, Bio-Serv
Supplier: Bio-Serv
K3262 K3261 K3261 K3262 K3365 K3365
89067-722EA 22.47 USD
89067-722 CA89067-722 89067-724 89067-726 CA89067-724 CA89067-726
Rodent Huts, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Rodent Huts are made of high-temp polycarbonate
The Fat Rat Hut's low-profile design fits into most standard shoebox rat cages. The larger design allows "oversize" rats an opportunity for shelter. Red tint blocks visible light, giving rats a sense of security.
They provide a comfortable hiding place. Transparent material allows for ease of visibility. Stacks easily for storage. Autoclavable. Cagewasher safe. Red.
Certifications: Certified (Contaminant Screened).