Formalin 10%, carbonate buffered (neutral), Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Methylaldehyde
, Formol
, Paraformaldehyde aqueous solution
, Formaldehyde
M-Clarity™ quality level = MQ300
R03379-86
R03379-86
R03379-82
R03379-82
EM-R03379-86EA
309
USD
CAR03379-82
CAR03379-86
EM-R03379-86
EM-R03379-82
Formalin 10%, carbonate buffered (neutral), Sigma-Aldrich®
Formalin
Formalin. Neutral 10%, Carbonate Buffered.
Formula:
CH₂O MW: 30.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 |
Specification Test Results
N/A | N/A |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...