BioFX® BCIP/NBT Blue One Component AP Membrane Substrate, Surmodics
Supplier: Surmodics
BCIB-0100-01 BCIB-1000-01
100359-210EA 50 USD
100359-210 100359-212
BioFX® BCIP/NBT Blue One Component AP Membrane Substrate, Surmodics
Assays ELISAs
The substrate is a ready to use, one-component formluation of 5-bromo-4-chloro-3-indolylphosphate and nitroblue tetrazolium for use in immunblotting or other membrane applications.
  • Ready to Use
  • 3 year shelf life
  • 1-Component
The BCIP/NBT substrate provides a dark blue precipitate upon reaction with alkaline phosphtase.
Certifications: ISO 13485:2016 & 9001:2015 Certified