Anti-ST8SIA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24918-1-AP-150UL
10155-354EA 462.47 USD
10155-354
Anti-ST8SIA1 Rabbit Polyclonal Antibody
Antibodies
The ST8SIA1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ST8SIA1. This antibody recognizes human antigen. The ST8SIA1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000 ; IHC 1 : 400

Type: Primary
Antigen: ST8SIA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR