Potassium-tert-butoxide ∼20% in THF for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Potassium tert-butanolatePotassium t-butoxidePotassium tert-butoxidePotassium tert-butylate

M-Clarity™ quality level = MQ200

8.43849.0250 8.43849.0250 8.43849.0050 8.43849.0050
EM8.43849.0250CS 2720.23 USD
EM8.43849.0250 EM8.43849.0050 CA8.43849.0050 CA8.43849.0250
Potassium-tert-butoxide ∼20% in THF for synthesis, Sigma-Aldrich®
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
MDL Number: MFCD00012162
CAS Number: 865-47-4
UN: 2920
ADR: 8 (3),I

Order Now

Specification Test Results

Solubility (20 °C) Risk of violent reaction.
Flash Point -21.5 °C (THF)
Density 0.914 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR