Trichloroacetic acid (TCA) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: TCAAceto-CaustinTrichloroethanoic acid

M-Clarity™ quality level = MQ200

8.22342.0250 8.22342.1000 8.22342.1000 8.22342.0025 8.22342.0025
EM8.22342.1000CS 1423.46 USD
EM8.22342.1000 EM8.22342.0250 EM8.22342.0025 CA8.22342.1000 CA8.22342.0025
Trichloroacetic acid (TCA) for synthesis, Sigma-Aldrich®
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
MW: 163.39 g/mol
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 1839
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/74/52/727452.jpg

Order Now

Specification Test Results

Solubility 1600 g/l (20 °C)
Melting Point 54 - 56 °C
Molar Mass 163.38 g/mol
Bulk Density 900 kg/m3
Boiling Point 197 °C
Vapor Pressure 1 hPa (20 °C)
Flash Point 110 °C
Density 1.63 g/cm3 (20 °C)
pH  - 1 (50 g/l, H2O, 20 °C)
Ignition Point 711 °C
Hill Formula C2HCl3O2
Chemical Formula CCl3COOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR