1-Docosanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Behenyl alcohol

M-Clarity™ quality level = MQ200

8.22047.0100 8.22047.0100
EM8.22047.0100CS 1317.37 USD
CA8.22047.0100 EM8.22047.0100
1-Docosanol for synthesis, Sigma-Aldrich®
1-Docosanol
Formula: H₃C(CH₂)₂₀CH₂OH
MW: 326.61 g/mol
MDL Number: MFCD00002939
CAS Number: 661-19-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 68 - 71 °C
Molar Mass 326.6 g/mol
Boiling Point 180 °C (0.3 hPa)
Synonyms Behenyl alcohol
Hill Formula C22H46O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR