Dapsone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Aminophenyl sulphone1-Amino-4-((4-aminophenyl)sulphonyl)benzene4,4'-sulfonyldianiline4-Aminophenyl sulfone4,4'-Diamino diphenyl sulfone4,4'-Sulfonyldianiline1-Amino-4-((4-aminophenyl)sulfonyl)benzene4,4'-SulphonyldianilineDDS4-[(4-Aminophenyl)sulphonyl]aniline4,4'-Diamino diphenyl sulphone4-[(4-Aminophenyl)sulfonyl]aniline4,4'-sulphonyldianiline

M-Clarity™ quality level = MQ200

8.21073.0250 8.21073.0005 8.21073.0250 8.21073.0005
EM8.21073.0250CS 2938 USD
CA8.21073.0005 EM8.21073.0250 CA8.21073.0250 EM8.21073.0005
Dapsone for synthesis, Sigma-Aldrich®
Dapsone
Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
MDL Number: MFCD00007887
CAS Number: 80-08-0

Order Now

Specification Test Results

Solubility 0.38 g/l (37 °C)
Melting Point 172 - 175 °C
Molar Mass 248.30 g/mol
Bulk Density 250 - 350 kg/m3
pH 5.5 - 7.5 (H2O, 20 °C) (saturated aqueous solution)
Synonyms DDS, 4,4'-Sulfonyldianiline, Bis(p-aminophenyl) sulfone
Hill Formula C12H12N2O2S
Chemical Formula [4-(NH2)C6H4]2SO2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR