Phenazine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.20973.0005
8.20973.0005
EM8.20973.0005CS
95865
USD
CA8.20973.0005
EM8.20973.0005
Phenazine for synthesis, Sigma-Aldrich®
Phenazine
Formula:
C₁₂H₈N₂ MW: 180.21 g/mol |
MDL Number:
MFCD00005023 CAS Number: 92-82-0 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 173 - 175 °C |
Molar Mass | 180.21 g/mol |
Boiling Point |
|
Synonyms | Dibenzopyrazine |
Hill Formula | C12H8N2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...