Benzalkonium chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
M-Clarity™ quality level = MQ200
8.21944.0100
8.21944.0500
EM8.21944.0100CS
750
USD
EM8.21944.0100
EM8.21944.0500
Benzalkonium chloride for synthesis, Sigma-Aldrich®
Benzalkonium chloride
Formula:
C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇) |
MDL Number:
MFCD00145757 CAS Number: 63449-41-2 UN: 1759 ADR: 8 III |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 37 - 41 °C |
Bulk Density | 800 kg/m3 |
Boiling Point | 115 °C |
pH | 6 - 8 (10 g/l, H2O, 20 °C) |
Synonyms | Benzyldimethylalkylammonium chloride |
Hill Formula | [RN(CH3)2CH2C6H5]Cl |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...