2,3-Dimercapto-1-propanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
2,3-Dimercaptopropanol
, Dithioglycerine
, Dimercaprol
, Dimercapto-2,3 propanol-1
, 2,3-Dimercaptopropane-1-ol
, 2,3-Dimercaptopropan-1-ol
, BAL
M-Clarity™ quality level = MQ200
8.18757.0025
8.18757.0025
8.18757.0005
8.18757.0005
EM8.18757.0005CS
224597
USD
EM8.18757.0005
EM8.18757.0025
CA8.18757.0005
CA8.18757.0025
2,3-Dimercapto-1-propanol for synthesis, Sigma-Aldrich®
2,3-Dimercapto-1-propanol
Formula:
C₃H₈OS₂ MW: 124.22 g/mol |
MDL Number:
MFCD00004864 CAS Number: 59-52-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Solubility | 87 g/l (20 °C) (slow decomposition) |
Molar Mass | 124.22 g/mol |
Boiling Point | 63 - 68 °C (0.27 hPa) |
Vapor Pressure | 7.4 hPa (100 °C) |
Flash Point |
|
Density | 1.24 g/cm3 (20 °C) |
pH | 5.0 - 6.5 (H2O, 20 °C) (saturated solution) |
Synonyms | Dithioglycerine, Dimercaprol, BAL |
Hill Formula | C3H8OS2 |
Chemical Formula | HOCH2CH(SH)CH2SH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...