Cyclohexyl isocyanide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18151.0010 8.18151.0010
EM8.18151.0010CS 163191 USD
CA8.18151.0010 EM8.18151.0010
Cyclohexyl isocyanide for synthesis, Sigma-Aldrich®
Cyclohexyl isocyanide
Formula: C₇H₁₁N
MDL Number: MFCD00003839
CAS Number: 931-53-3
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Molar Mass 109.17 g/mol
Boiling Point 173 - 176 °C (1013 hPa)
Flash Point 77 °C
Refractive Index 1.4502 (20 °C, 589 nm)
Density 0.89 g/cm3 (20 °C)
Synonyms Cyclohexyl isonitrile
Hill Formula C7H11N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR