Benzalkonium chloride 50% water zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride

MQ200

8.14858.1000 8.14858.5000
EM8.14858.1000CS 500 USD
EM8.14858.1000 EM8.14858.5000
Benzalkonium chloride 50% water zS, Sigma-Aldrich®
Benzalkonium chloride
Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
MDL Number: MFCD00145757
CAS Number: 63449-41-2
UN: 1760
ADR: 8 III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -5 °C
Boiling Point 100 °C
Flash Point 160 - 170 °C
Density 0.978 - 0.988 g/cm3 (20 °C)
pH 5 - 7 (10 g/l, H2O, 20 °C)
Synonyms Benzalkonium chloride (50% aqueous solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR