Ethyl oleate mixture of esters stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14762.0250 8.14762.1000
EM8.14762.0250CS 689 USD
EM8.14762.0250 EM8.14762.1000
Ethyl oleate mixture of esters stabilized for synthesis, Sigma-Aldrich®
Ethyl oleate

The material is stabilised with Butylhydroxyanisole (BHA). At time of analytical testing a content of >10 ppm stabiliser was determined.

Formula: C₂₀H₃₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -32 °C
Boiling Point 168 °C (1.3 hPa)
Flash Point 110 °C
Density 0.87 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR