Platinum 10% on activated carbon hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Platinium
M-Clarity™ quality level = MQ200
8.07339.0005
8.07339.0005
8.07339.0001
8.07339.0001
EM8.07339.0001CS
49578.02
USD
EM8.07339.0001
EM8.07339.0005
CA8.07339.0001
CA8.07339.0005
Platinum 10% on activated carbon hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Platinum
Formula:
Pt MW: 195.08 g/mol |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 UN: 3088 ADR: 4.2,III |
Specification Test Results
Solubility | (20 °C) insoluble |
Bulk Density | 250 kg/m3 |
pH | 2 - 10 (50 g/l, H2O, 20 °C) |
Ignition Point | 600 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...