4-Amino-4H-1,2,4-triazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-Amino-1,2,4-triazole4H-1,2,4-Triazol-4-amine1,2,4-Triazol-4-ylamine

M-Clarity™ quality level = MQ200

8.01309.0025 8.01309.0025
EM8.01309.0025CS 7576 USD
CA8.01309.0025 EM8.01309.0025
4-Amino-4H-1,2,4-triazole for synthesis, Sigma-Aldrich®
4-Amino-4H-1,2,4-triazole
Formula: C₂H₄N₄
MW: 84.08 g/mol
MDL Number: MFCD00003099
CAS Number: 584-13-4

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 83 - 86 °C
Molar Mass 84.08 g/mol
pH  - 7 (H2O, 20 °C)
Synonyms 4-Amino-4H-1,2,4-triazole
Hill Formula C2H4N4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR