Glutaric acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,5-Pentanedioic acid
M-Clarity™ quality level = MQ200
8.00295.0100
8.00295.0250
8.00295.1000
EM8.00295.0100CS
6348
USD
EM8.00295.0100
EM8.00295.0250
EM8.00295.1000
Glutaric acid for synthesis, Sigma-Aldrich®
Glutaric acid
Formula:
C₅H₈O₄ MW: 132.12 g/mol Boiling Pt: 302 Melting Pt: 98 |
MDL Number:
MFCD00004410 CAS Number: 110-94-1 UN: 3261 ADR: 8 II Merck Index: 13,04486 |
Specification Test Results
Solubility | 640 g/l (20 °C) |
Melting Point | 98 °C |
Molar Mass | 132.11 g/mol |
Boiling Point | 302 - 304 °C (1013 hPa) (slow decomposition) |
Vapor Pressure | 0.022 hPa (18.5 °C) |
Density | 1.43 g/cm3 (15 °C) |
Synonyms | Pentanedioic acid |
Hill Formula | C5H8O4 |
Chemical Formula | HOOC(CH2)3COOH |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...