Anti-FUT7 Rabbit Polyclonal Antibody

Supplier: Proteintech
18197-1-AP-150UL
10154-806EA 462.47 USD
10154-806
Anti-FUT7 Rabbit Polyclonal Antibody
Antibodies
The FUT7 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FUT7. This antibody recognizes human,mouse,rat antigen. The FUT7 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 200

Type: Primary
Antigen: FUT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR