Anti-KCNN4 Mouse Monoclonal Antibody [clone: 2A6G5]

Supplier: Proteintech
60276-1-IG-150UL
10155-876EA 462.47 USD
10155-876
Anti-KCNN4 Mouse Monoclonal Antibody [clone: 2A6G5]
Antibodies
The KCNN4 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human KCNN4. This antibody recognizes human, mouse, rat antigen. The KCNN4 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: KCNN4
Clonality: Monoclonal
Clone: 2A6G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR