StabilGuard® Choice Stabilizer, Surmodics
Supplier: Surmodics
StabilGuard®
SG02-1000
102466-316EA 258.46 USD
102466-316
StabilGuard® Choice Stabilizer, Surmodics
Microarray Reagents
StabilGuard® Choice Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilGuard Choice stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.
  • Protein-Free
  • 3 year shelf life
  • Increased stability
  • Reduces non-specific binding
StabilGuard Choice Microarray Stabilizer (BSA-Free) improves the stability of proteins covalently bound to microspheres stored in solution and array proteins stored in the dried state, without the use of BSA protein.
Certifications: ISO 13485:2016 & 9001:2015 Certified