ActivArmr® 52-547 General purpose gloves, Ansell
Supplier: Ansell Healthcare
207481 207480
32890-232CS 1661.28 USD
32890-232 47742-590
ActivArmr® 52-547 General purpose gloves, Ansell
Gloves
With nitrile-impregnated coating on a jersey liner, these gloves supersede the classic leather general-purpose glove in medium-duty applications.
  • Great flexibility and comfort
  • Excellent dry grip for quicker, easier handling
  • Resist shrinkage in machine washing
  • Long-lasting durability ensures economical reuse
These nitrile coated gloves are more flexible, more comfortable, and far more hard-wearing.
Use in the agriculture and viticulture, automotive and transportation, building and construction, chemical, glass, and metal fabrication industries. Uses include assembling metal parts and components; assembly and finishing; gas and electricity supply; general purpose, offshore; handling light castings and metal parts; injection molding; handling cold components; maintenance; mining; operating machinery; and warehouse work.
The high-performance nitrile impregnation offers superior resistance to abrasion and good protection against snags, cuts and puncture. Effectively repels grease, oil and dirt. The soft, flexible jersey lining offers a close, comfortable fit while safely cushioning the hand. Absorbs perspiration, keeping the hand dry and comfortable for longer. Gloves provide excellent dry grip and make handling quicker and easier. Safely machine-washable: will not shrink, stiffen or lose shape. More hygienic and longer lasting, they can safely and economically be reused.
Certifications: ANSI level 1 cut protection; ANSI Level 3 Abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.