Edge® 40-400 Palm-Coated Nitrile Gloves, Ansell
Supplier: Ansell Healthcare
218710 218711 218712
32890-168CS 1336.44 USD
32890-168 32890-175 32890-170 32890-172
Edge® 40-400 Palm-Coated Nitrile Gloves, Ansell
Gloves
Gloves mold to the hand, ensuring improved movement and dexterity, and in slightly oily applications, the porous nitrile coating helps channel oil away from the glove surface for a firmer grip.
  • Ergonomically designed
  • Lightweight and flexible
  • Porous coating channels oil away
Get a protective edge over ordinary cotton and leather gloves.
Use in the automotive and transportation, building and construction, machinery and equipment, and metal fabrication industries. Uses include assembling metal parts and components; grinding and fettling; maintenance; metal fabrication; operating punches and presses; shipping and receiving; and spot welding.
Gloves can outwear split leather. Ergonomic, light, flexible, and tough, these gloves are the ideal combination for quick and accurate handling.
Certifications: ANSI level 1 cut protection; ANSI Level 3 Abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.