Microwave Oven, Argos Technologies
Supplier: Argos Technologies
24406-40
10147-786EA 1108.46 USD
10147-786
Microwave Oven, Argos Technologies
Ovens Microwave Ovens
The compact design makes this .7 cu foot microwave oven ideal for the crowded lab as it occupies minimal counter space
Its 700 watts of power prepares samples rapidly without overheating. The control panel is easy to read and operate, and can be locked to prevent changes to the program. The rapid heat feature sets the oven at high with a single touch. Internal glass turntable rotates during operation for even heat distribution.
Certifications: UL Listed.