GHS Labels "Acute Toxic Picto", Brady

Supplier: Brady Worldwide
121193 121208 121209
89234-544EA 45.14 USD
89234-544 89234-574 89234-576
GHS Labels "Acute Toxic Picto", Brady
Labels
Pre-printed GHS pictogram die cut labels on a card.

  • Color: Black/red on white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR