Fibroblast Growth Medium 3 for Cardiac Fibroblasts, PromoCell
Supplier: Promocell, Inc.
PromoCell Fibroblast Growth Medium 3 has been developed for the in vitro cultivation of adult Human Cardiac Fibroblasts (HCF)
The medium is optimized for primary human cells but can also be used for porcine cardiac fibroblasts
Ordering information: PromoCell Fibroblast Growth Medium 3 is available as Medium (Ready-touse) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.