Anti-IL4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MP4-25D2]

Supplier: Tonbo Biosciences
50-7048-T025 50-7048-T100
10208-722EA 66 USD
10208-722 10208-724
Anti-IL4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MP4-25D2]
Antibodies
The MP4-25D2 antibody is specific for human IL-4, a pleiotropic cytokine that Th2 cell differentiation, B cell proliferation and differentiation, and B cell class switching to IgE. MP4-25D2 is a neutralizing antibody and is reported to cross react with rhesus monkey IL-4.

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human, Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR