Close-it®, Tapes, Bürkle
Supplier: BURKLE INC
5303-0017 5303-2019
10216-988PK 140.61 USD
10216-988 10217-008
Close-it®, Tapes, Bürkle
Tapes
With its extremely strong adhesive strength, close-it® tapes adhere to a range of different surfaces, where the glue effect of normal adhesive tape quickly reaches its maximum capacity.
  • Customisable
  • White with tear-off lines
  • Available in the classic close-it® design, as well as close-it® tape food, with a food-compatible adhesive
  • Can be labelled with permanent ink and provides gas-proof seal
Standard close-it® and close-it® food control seals are also available on a continuous roll. This offers the flexibility to provide a small length of tape to sealMore Product Information