Labels, Diversified Biotech
Supplier: Diversified Biotech
WetGrip
GRLA-1050 GRLA-2016 GRDT-3000 GRDT-6000 GRDT-1000
490006-752EA 66 USD
490006-752 490006-756 490006-754 490014-308 490014-306
Labels, Diversified Biotech
Labels
WetGrip labels are suitable for moist, frost covered tubes.
  • Compatible with centrifuge rotors
  • Resistant to chemicals, solvents and temperature up to –80 °C
It is available in laser sheets for printing many labels at a time, direct thermal for printing few labels at a time and labels on rolls for handwriting labels.