Food Control Seal Labels, Bürkle
Supplier: Bürkle
Close-it®
5303-3020 5303-3023 5303-3022 5303-3021 5303-2022 5303-2020 5303-3017 5303-2023 5303-2017 5303-2021 5303-3018
10217-028PK 202.1 USD
10217-028 10217-018 10217-026 10217-004 10217-016 10217-020 10217-010 10217-030 10217-024 10217-014 10217-012
Food Control Seal Labels, Bürkle
Labels
A control seal with an adhesive approved for use with foodstuffs. The strength of the adhesive is almost as powerful as that of the original. Just like the original, close-it® food has a barrier layer of aluminium foil. This means that close-it® food is suitable for sampling foodstuffs, pharmaceuticals, cosmetics, food additives, even frozen goods where cleanliness is particularly important.
  • Adheres very firmly
  • Can be written on
  • According to FDA guidelines
  • Particularly suited for the food and pharma industry
Packaging: Rolls contain 500 (95×95 mm) or 250 (150×150 mm) labels.