Anti-TNFSF4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RM134L]

Supplier: Tonbo Biosciences
20-5905-U025 20-5905-U100
10210-118EA 66 USD
10210-118 10210-120
Anti-TNFSF4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RM134L]
Antibodies
The RM134L antibody recognizes CD252, also known as OX-40 Ligand or CD134 Ligand, a member of the TNF superfamily that is present on the surface of antigen presenting cells and activated B lymphocytes. The OX-40 Ligand interacts with OX-40 (CD134) which is expressed primarily on activated T cells. This costimulatory interaction leads to increased proliferation and IL-2 production responses of activated T cells, and at the same time enhances proliferation and immunoglobulin secretion by activated B cells.

The RM134L antibody is useful for flow cytometric detection of CD252 on stimulated mouse splenocytes. It has also been reported to block the costimulatory activity of OX-40 Ligand. Please choose the appropriate format for each application.

Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: RM134L
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR