Coomassie® brilliant blue R-250, dark powder for electrophoresis
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Brillant blue R
, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-benzenemethanaminium inner salt, sodium salt (1:1)
, Brilliant Blue R250
, Brilliant indocyanine 6B
, Brilliant Blue R
, Serva Blue R
, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1)
, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt
, Coomassie Blue R 250
, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt
, Acid Blue 83
, Brilliant Cyanine 6B
Coomassie® Brilliant Blue R-250 is a fast acting, sensitive dye, which can be used on SDS gels, IEF gels, and standard gels.
- For Detection of Protein Bands Following Electrophoresis
Brilliant Blue R can be used for staining proteins on polyacrylamide electrophoresis gels. Improvement of sensitivity and background have been accomplished using Brilliant Blue dyes in a colloidal form. Other applications for brilliant Blue R include use in detection of lipids in thin layer chromatography, determination of the CMC of detergents, and detection of proteins in solution, as well as on cellulose acetate plates.
Soluble in water (10 mg/ml), 20 mg/ml in methyl cellosolve, ethanol (10 mg/ml), methanol.
Caution: Research Use Only (RUO).
Formula:
C₄₅H₄₄N₃NaO₇S₂ MW: 825.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041762 CAS Number: 6104-59-2 |
Specification Test Results
Identity Test | Passes |
λmax (ethanol) | 585 ± 5 nm |
Extinction Coefficient | E1%(587nm) = 785 (methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...