Sodium hydrogen carbonate 99,5, powder_white

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium bicarbonate

02195497.5 0219549701 0219549705 0219549703
IC195497.5EA 65.88 USD
IC195497.5 IC19549701 IC19549705 76177-270
Sodium hydrogen carbonate 99,5, powder_white
Sodium hydrogen carbonate

Soluble in water (100 mg/mL).

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 (dec., lit.)
Density: 2.159 (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f3a3db0ff523037396b24e8e3130f012274f40a&vid=3f3a3db0ff523037396b24e8e3130f012274f40a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99.5%
pH (0.1M aq soln) 8.1 - 8.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR