Toluene-α-sulfonyl fluoride, white to off white needle crystals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulfonyl fluoride, α-Toluenesulphonyl fluoride, Phenylmethanesulfonyl fluoride, Phenylmethylsulfonyl fluoride

02195381.2 0219538101 0219538105 0219538125 0219538180
IC195381.2EA 62.3 USD
IC195381.2 IC19538101 IC19538105 IC19538125 IC19538180
Toluene-α-sulfonyl fluoride, white to off white needle crystals
Toluene-α-sulphonyl fluoride

Phenylmethylsulfonyl fluoride is an irreversible inhibitor of serine proteases - inhibits trypsin, chymotrypsin and protease. Inhibits mammalian acetyl cholinesterase, but not those from fish sources.


  • Presentation: White to off white needle crystals
  • Causes sulfonylation of the active-site serine residue
  • A less toxic substitute for the extremely toxic diisopropylfluorophosphate


Phenylmethylsulfonyl fluoride inhibits serine proteases such as trypsin and chymotrypsin. Also inhibits cysteine proteases (reversible by reduced thiols) and mammalian acetylcholinesterase.
Administration of PMSF produces analgesia unrelated to its anticholinesterase effect, and prolongs the analagesic effect of centrally administered β-endorphin.


Soluble in methanol and ethanol


Phenylmethylsulfonyl fluoride is an irreversible inhibitor of serine proteases - inhibits trypsin, chymotrypsin and protease. Causes sulfonylation of the active-site serine residue. A less toxic substitute for the extremely toxic diisopropylfluorophosphate. Inhibits mammalian acetyl cholinesterase, but not those from fish sources. Recommended concentration is 15 to 175 ug/mL.

Formula: C₇H₇FO₂S
Melting Pt: 89…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007424
CAS Number: 329-98-6
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75d4b1d585a1f8fa4431f9f6020e45c800861488&vid=75d4b1d585a1f8fa4431f9f6020e45c800861488&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR