n-Hexadecane ≥99%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Hexadecane, Cetane

0219521880 0219521890 0219521891
IC19521880EA 93.54 USD
IC19521880 IC19521890 IC19521891
n-Hexadecane ≥99%, liquid
n-Hexadecane

Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.


Soluble in ether; insoluble in water.


Transition temp.- solidification point 17.5 - 18.5 °C (Lit.)

Formula: H₃C(CH₂)₁₄CH₃
MW: 226.45 g/mol
Boiling Pt: 287
Melting Pt: 18
Flash Pt: 135
MDL Number: MFCD00008998
CAS Number: 544-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32532c13634f211b15a7db59a745c30dd17fb2e2&vid=32532c13634f211b15a7db59a745c30dd17fb2e2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR