Calcium chloride dihydrate ∼99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Calcium dichlorideCalcium Chloride Dihydrate

0219508890
IC19508890EA 114.54 USD
IC19508890
Calcium chloride dihydrate ∼99%, white crystalline powder
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
MDL Number: MFCD00149613
CAS Number: 10035-04-8

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
pH (5% aq soln) 4 - 9.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR