Rifamycin SV sodium

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione sodium salt, Rifamycin sodium

02191516.2 0219151601 0219151605
IC191516.2EA 66.17 USD
IC191516.2 IC19151601 IC19151605
Rifamycin SV sodium
Rifamycin SV sodium

Rifamycin SV is a broad-spectrum semisynthetic antimicrobial agent of the rifamycin group.


  • Orange powder


Rifamycin SV inhibits selective (E. coli, B. subtilis) bacterial DNA-dependent RNA polymerase by binding to the polymerase β-subunit, a mechanism similar to rifabutin. It acts as a selective cytochrome P450 3A4 inducer. It has limited oral absorption that is active against Gram-positive bacteria and is moderately active against Gram-negative organisms.


Soluble in Ethanol (50 mg/mL)

Formula: C₃₇H₄₆NNaO₁₂
Storage Temperature: Refrigerator
MDL Number: MFCD00056847
CAS Number: 14897-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR