Potassium iodide, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Potassium Iodide is known as Potassium Salt of Hydriodic Acid [HI], available in white crystalline powder form. It is a mild reducing agent and can be easily oxidized to Iodine by powerful oxidizing agent like chlorine. With elemental iodine it forms I3 (tri iodide), the salts of tri iodide are highly water soluble.
Readily soluble in water.
Potassium iodide is used in organic synthesis for the preparation of Aryl iodides and Iodo alkanes. It is used in biochemistry, often as a fluorescence quenching agent which acts by collisional quenching of fluorescent substances by the iodide ion. It is also used in extraction of single stranded DNA.
Potassium Iodide is known as Potassium Salt of Hydriodic Acid [HI], available in white crystalline powder form. It is a mild reducing agent and can be easily oxidized to Iodine by powerful oxidizing agent like chlorine. With elemental iodine it forms I3 (tri iodide), the salts of tri iodide are highly water soluble.
Formula:
Kl Melting Pt: 681 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 |
Specification Test Results
Identity Test | Passes |
pH (5% aq soln) | 6 - 9.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...