Succinic acid ≥99%, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 1,4-Butanedioic acid

0219144980
IC19144980EA 75.1 USD
IC19144980
Succinic acid ≥99%, powder ACS
Succinic acid

Succinic acid is a dicarboxylic acid that occurs in nature in such organisms as fungi and lichens. In eukaryotes, succiniate, the anion of succinic acid, is an intermediate in the citric acid cycle, being formed from succinyl CoA and being converted to fumarate. Bacteria and plants can produce succinate from acetate or acetyl CoA, in the glyoxylate cycle.


Succinic acid has been used in large scale applications including the manufacture of lacquers, dyes, and esters for perfumes. In polymer research, succinic acid has been utilized to prepare biocompatible hybrid dendritic-linear polyester-ethers. A study of the co-crystallization of cis-itraconazole with various 1,4-dicarboxylic acids, including succinic acid, has been reported. Succinic acid has been used as a matrix in infrared (IR) MALDI analytical methods. An analytical study of various low molecular weight organic acids, including succinic acid, using capillary zone electrophoresis-electrospray ionization mass spectrometry has been publishe


Chloride ≤0.001% ; Phosphate ≤0.001% ; Sulfate ≤0.003% ; Nitrogen Compounds ≤0.001%


Soluble in water (100 mg/mL - clear, colorless solution), alcohol (54 mg/mL), methanol (158 mg/mL), acetone (27 mg/mL), glycerol (50 mg/mL); practically insoluble in benzene, carbon disulfide, carbon tetrachloride, petroleum and ether (8.8 mg/mL).

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
UN: 3261
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/01/38/2300138.jpg

Order Now

Specification Test Results

Assay ≥99%
Melting range 185.0-191.0°C (w/i 2°C)
Insoluble matter 0,01%
Residue on Ignition 0,02%
Chloride ≤0.001%
Phosphate ≤0.001%
Sulphate ≤0.003%
Nitrogen compounds ≤0.001%
Heavy Metals ≤5 ppm
Iron ≤5 ppm
FTIR Conforms to standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR