Sodium phosphate dibasic, anhydrous ≥99.0%, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: di-Sodium hydrogen phosphate anhydrousSodium phosphate dibasicsec-Sodium phosphateDisodium phosphatedi-Sodium hydrogen orthophosphateSodium hydrogen phosphate

02191440.1 02191440.5 0219144005 0219144001
IC191440.5EA 91.48 USD
IC191440.5 IC19144001 IC191440.1 IC19144005
Sodium phosphate dibasic, anhydrous ≥99.0%, white crystalline powder ACS
di-Sodium hydrogen phosphate

Sodium dibasic anhydrous is used as a sequestrant, emulsifier and buffer in foods, as mordant in dyeing for weighting silk, in tanning, in manufacturing of enamels, ceramics, detergents, boiler compounds; as fireproofing agent in soldering and brazing instead of borax; as reagent and buffer in analytical chemistry, cathartic.,laxative.


It a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.


Soluble in water, but more soluble in hot water; insoluble in alcohol, ethanol.


Caution: Research Use Only (RUO).

Formula: Na₂HPO₄
MW: 141.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4

Order Now

Specification Test Results

Assay ≥99.0%
pH (5% aq soln) 8,7 - 9,3
Insoluble Matter ≤0,01%
Loss on Drying (105 °C) ≤0,2%
Chloride (Cl) ≤0,002%
Sulphate (SO4) ≤0,005%
Heavy Metals ≤0,001%
pH ≤0,002%
pka Iron (Fe)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR