di-Potassium hydrogen phosphate, anhydrous ≥98%, white crystalline powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
di-Potassium hydrogen orthophosphate
, Dibasic Potassium Phosphate
, Potassium phosphate dibasic
, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate)
, Potassium hydrogen phosphate
, Dipotassium hydrogen phosphate
Potassium phosphate is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.
Soluble in water; Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Caution: Research Use Only (RUO).
Formula:
K₂HPO₄ MW: 174.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011383 CAS Number: 7758-11-4 |
Specification Test Results
Assay | ≥98% |
pH (5% aq soln) | 8,5 - 9,6 |
Insoluble Matter | ≤0,01% |
Loss on Drying | ≤1,0% |
Chloride (Cl) | ≤0,003% |
Nitrogen (as N) | ≤0,001% |
Sulfate (SO₄) | ≤0,005% |
Heavy Metals (as Pb) | ≤5 ppm |
Iron (Fe) | ≤0,001% |
Sodium (Na) | ≤0,05% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...