Potassium nitrate ≥99.0%, crystalline powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Nitrate of Potash
0219142801
02191428.5
02191428.1
0219142805
IC19142801EA
198.22
USD
IC19142801
IC191428.1
IC19142805
IC191428.5
Potassium nitrate ≥99.0%, crystalline powder ACS
Potassium nitrate
Soluble in water (133 mg/mL), yielding a clear, colorless solution. soluble in glycerol; insoluble in absolute alcohol.
Chloride (Cl): ≤0.002%; Iodate (IO3): ≤5 ppm; Nitrite (NO2): ≤0.001%; Phosphate (PO4): ≤5 ppm; Sulfate (SO4): ≤0.003%; Calcium (Ca): ≤0.005%; Magnesium (Mg): ≤0.002%; Sodium (Na): ≤0.005%
Formula:
KNO₃ MW: 101.1 g/mol Melting Pt: 334 °C |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 UN: 1486 ADR: 5.1,III |
Specification Test Results
Synonyms | Niter; Nitre; Nitric Acid Potassium Salt; Saltpeter; Vicknite |
Form | White Crystalline Powder |
Storage | Store at Room Temperature (15-30 °C). |
Assay | ≥99.0% |
Heavy Metals | ≤5 ppm(as Pb); Iron (Fe): ≤3 ppm |
Insoluble Matter | ≤0.005% |
pH | 4.5 - 8.5 (5% aq soln) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...