Ammonium acetate, anhydrous ≥97%, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219140480 0219140490 0219140491
IC19140480EA 78.11 USD
IC19140480 IC19140490 IC19140491
Ammonium acetate, anhydrous ≥97%, powder ACS
Ammonium acetate

Chloride (Cl) ≤5 ppm; Nitrate (NO3) 0.001%; Sulfate (SO4) ≤0.001%; Iron (Fe) ≤5 ppm

Formula: H₃CCOONH₄
MDL Number: MFCD00013066
CAS Number: 631-61-8

Order Now

Specification Test Results

Synonyms Acetic acid ammonium salt
Form White Crystalline Powder
Storage Store at Room Temperature (15-30 °C). Store Desiccated.
Assay ≥97%
Heavy Metals ≤5 ppm
Insoluble Matter ≤0.005%
pH 6.7 - 7.3 (5% aq soln)
Residue on Ignition ≤0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR